Assay Details
| Gene Symbol: LOC4326812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766814.2 | XP_015622300.1 | 19 - 20 | 3624 | 94 | |
| XM_015766815.2 | XP_015622301.1 | 18 - 19 | 4577 | 94 | ||
| GenBank mRNA | AK106603.1 | - | 6 - 7 | 1186 | 94 | |
| AY332479.1 | - | 19 - 20 | 3453 | 94 | ||
More Information
Set Membership: |
|

LOC4326812
Genomic Map