Assay Details
| Gene Symbol: LOC4326812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766814.2 | XP_015622300.1 | 5 - 6 | 998 | 104 | |
| XM_015766815.2 | XP_015622301.1 | 4 - 5 | 1951 | 104 | ||
| GenBank mRNA | AY332479.1 | - | 5 - 6 | 827 | 104 | |
More Information
Set Membership: |
|

LOC4326812
Genomic Map