Assay Details
| Gene Symbol: LOC4326283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772035.2 | XP_015627521.1 | 2 - 3 | 975 | 103 | |
| GenBank mRNA | AK120976.1 | - | 2 - 3 | 904 | 103 | |
| Y11189.1 | - | 2 - 3 | 134 | 103 | ||
More Information
Set Membership: |
|

LOC4326283
Genomic Map