Assay Details
| Gene Symbol: LOC4326283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772035.2 | XP_015627521.1 | 5 - 6 | 1246 | 134 | |
| GenBank mRNA | AK120976.1 | - | 5 - 6 | 1175 | 134 | |
| Y11189.1 | - | 5 - 6 | 405 | 134 | ||
More Information
Set Membership: |
|

LOC4326283
Genomic Map