Assay Details
| Gene Symbol: LOC4326027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765419.2 | XP_015620905.1 | 2 - 3 | 924 | 79 | |
| GenBank mRNA | AK060770.1 | - | 1 - 2 | 566 | 79 | |
| AK065117.1 | - | 2 - 3 | 929 | 79 | ||
| AK121916.1 | - | 2 - 3 | 927 | 79 | ||
More Information
Set Membership: |
|

LOC4326027
Genomic Map