Assay Details
| Gene Symbol: LOC4326027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765419.2 | XP_015620905.1 | 4 - 5 | 1140 | 75 | |
| GenBank mRNA | AK060770.1 | - | 3 - 4 | 782 | 75 | |
| AK065117.1 | - | 4 - 5 | 1145 | 75 | ||
| AK121916.1 | - | 4 - 5 | 1143 | 75 | ||
More Information
Set Membership: |
|

LOC4326027
Genomic Map