Assay Details
| Gene Symbol: LOC4326194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766868.2 | XP_015622354.1 | 11 - 12 | 1048 | 82 | |
| GenBank mRNA | AK062019.1 | - | 11 - 12 | 958 | 82 | |
| AK066233.1 | - | 11 - 12 | 1031 | 82 | ||
More Information
Set Membership: |
|

LOC4326194
Genomic Map