Assay Details
| Gene Symbol: LOC4326194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766868.2 | XP_015622354.1 | 1 - 2 | 156 | 56 | |
| GenBank mRNA | AK062019.1 | - | 1 - 2 | 66 | 56 | |
| AK066233.1 | - | 1 - 2 | 139 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015766868.2 transcripts.
Set Membership: |
|

LOC4326194
Genomic Map