Assay Details
| Gene Symbol: LOC4324196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766712.2 | XP_015622198.1 | 1 - 2 | 476 | 101 | |
| GenBank mRNA | AK066182.1 | - | 1 - 2 | 395 | 101 | |
| AK072015.1 | - | 1 - 2 | 408 | 101 | ||
More Information
Set Membership: |
|

LOC4324196
Genomic Map