Assay Details
| Gene Symbol: LOC4324196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766712.2 | XP_015622198.1 | 4 - 5 | 999 | 152 | |
| GenBank mRNA | AK066182.1 | - | 4 - 5 | 918 | 152 | |
| AK072015.1 | - | 4 - 5 | 931 | 152 | ||
More Information
Set Membership: |
|

LOC4324196
Genomic Map