Assay Details
| Gene Symbol: LOC4324196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766712.2 | XP_015622198.1 | 9 - 10 | 1614 | 82 | |
| GenBank mRNA | AK066182.1 | - | 9 - 10 | 1533 | 82 | |
| AK072015.1 | - | 9 - 10 | 1546 | 82 | ||
More Information
Set Membership: |
|

LOC4324196
Genomic Map