Assay Details
| Gene Symbol: LOC4326611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767799.2 | XP_015623285.1 | 1 | 63 | 57 | |
| GenBank mRNA | AK064632.1 | - | 1 | 57 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015767799.2 transcripts.
Set Membership: |
|

LOC4326611
Genomic Map