Assay Details
| Gene Symbol: LOC4326555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766979.2 | XP_015622465.1 | 3 - 4 | 578 | 84 | |
| GenBank mRNA | AK243497.1 | - | 3 - 4 | 537 | 84 | |
More Information
Set Membership: |
|

LOC4326555
Genomic Map