Assay Details
| Gene Symbol: LOC4325898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760926.2 | XP_015616412.1 | 15 - 16 | 5100 | 103 | |
| XM_015760935.2 | XP_015616421.1 | 13 - 14 | 3654 | 103 | ||
| XM_015760939.1 | XP_015616425.1 | 13 - 14 | 3732 | 103 | ||
| GenBank mRNA | AK107129.1 | - | 1 - 2 | 75 | 103 | |
| HQ858814.1 | - | 1 - 2 | 72 | 103 | ||
More Information
Set Membership: |
|

LOC4325898
Genomic Map