Assay Details
| Gene Symbol: LOC4325510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779805.2 | XP_015635291.1 | 2 - 3 | 866 | 53 | |
| GenBank mRNA | AK100701.1 | - | 2 - 3 | 868 | 53 | |
More Information
Set Membership: |
|

LOC4325510
Genomic Map