Assay Details
| Gene Symbol: LOC4325510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779805.2 | XP_015635291.1 | 3 - 4 | 1023 | 105 | |
| GenBank mRNA | AK100701.1 | - | 3 - 4 | 1025 | 105 | |
More Information
Set Membership: |
|

LOC4325510
Genomic Map