Assay Details
| Gene Symbol: LOC4325575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794134.1 | XP_015649620.1 | 3 - 4 | 748 | 95 | |
| GenBank mRNA | AK070307.1 | - | 3 - 4 | 710 | 95 | |
| AK120130.1 | - | 1 - 2 | 83 | 95 | ||
More Information
Set Membership: |
|

LOC4325575
Genomic Map