Assay Details
| Gene Symbol: LOC4325575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794134.1 | XP_015649620.1 | 4 - 5 | 866 | 66 | |
| GenBank mRNA | AK065152.1 | - | 2 - 3 | 1723 | 66 | |
| AK070307.1 | - | 4 - 5 | 828 | 66 | ||
| AK120130.1 | - | 2 - 3 | 201 | 66 | ||
More Information
Set Membership: |
|

LOC4325575
Genomic Map