Assay Details
| Gene Symbol: LOC4325575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794134.1 | XP_015649620.1 | 9 - 10 | 2236 | 75 | |
| GenBank mRNA | AK065152.1 | - | 7 - 8 | 3093 | 75 | |
More Information
Set Membership: |
|

LOC4325575
Genomic Map