Assay Details
| Gene Symbol: LOC4325591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778082.2 | XP_015633568.1 | 2 | 662 | 101 | |
| GenBank mRNA | AK111476.1 | - | 1 | 549 | 101 | |
More Information
Set Membership: |
|

LOC4325591
Genomic Map