Assay Details
| Gene Symbol: LOC9270029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761424.2 | XP_015616910.1 | 1 - 2 | 553 | 74 | |
| XM_015761432.2 | XP_015616918.1 | 1 - 2 | 553 | 74 | ||
| GenBank mRNA | AK072034.1 | - | 1 - 2 | 541 | 74 | |
| AK101627.1 | - | 1 - 2 | 539 | 74 | ||
More Information
Set Membership: |
|

LOC9270029
Genomic Map