Assay Details
| Gene Symbol: LOC4325619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773108.2 | 7 - 8 | 1387 | 96 | ||
| GenBank mRNA | AK070142.1 | - | 7 - 8 | 1391 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4325619| Gene Symbol: LOC4325619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773108.2 | 7 - 8 | 1387 | 96 | ||
| GenBank mRNA | AK070142.1 | - | 7 - 8 | 1391 | 96 | |
Set Membership: |
|