Assay Details
| Gene Symbol: LOC4325502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756470.2 | XP_015611956.1 | 2 - 3 | 632 | 89 | |
| GenBank mRNA | AK121269.1 | - | 2 - 3 | 462 | 89 | |
| AK122168.1 | - | 2 - 3 | 462 | 89 | ||
More Information
Set Membership: |
|

LOC4325502
Genomic Map