Assay Details
| Gene Symbol: LOC4327535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765657.2 | XP_015621143.1 | 2 - 3 | 551 | 84 | |
| GenBank mRNA | AK061770.1 | - | 2 - 3 | 553 | 84 | |
| AK072206.1 | - | 2 - 3 | 416 | 84 | ||
| AK121632.1 | - | 2 - 3 | 416 | 84 | ||
| BT129032.1 | - | 2 - 3 | 547 | 84 | ||
| BT129753.1 | - | 2 - 3 | 410 | 84 | ||
| BT131286.1 | - | 2 - 3 | 414 | 84 | ||
| KM982768.1 | - | 2 - 3 | 517 | 84 | ||
| Gene Symbol: LOC9271026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK241277.1 | - | 416 | 84 | ||
More Information
Set Membership: |
|

LOC4327535
Genomic Map