Assay Details
| Gene Symbol: LOC4327486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761291.1 | XP_015616777.1 | 29 - 30 | 3402 | 116 | |
| XM_015761295.1 | XP_015616781.1 | 29 - 30 | 3355 | 116 | ||
| GenBank mRNA | AK121331.1 | - | 29 - 30 | 3356 | 116 | |
More Information
Set Membership: |
|

LOC4327486
Genomic Map