Assay Details
| Gene Symbol: LOC4327502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770626.2 | XP_015626112.1 | 6 - 7 | 1954 | 78 | |
| GenBank mRNA | AK100245.1 | - | 6 - 7 | 1791 | 78 | |
More Information
Set Membership: |
|

LOC4327502
Genomic Map