Assay Details
| Gene Symbol: LOC4324742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765401.2 | XP_015620887.1 | 3 - 4 | 737 | 93 | |
| GenBank mRNA | AK072680.1 | - | 3 - 4 | 681 | 93 | |
More Information
Set Membership: |
|

LOC4324742
Genomic Map