Assay Details
| Gene Symbol: LOC4325074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786008.2 | XP_015641494.1 | 1 - 2 | 250 | 102 | |
| GenBank mRNA | AK060459.1 | - | 1 - 2 | 192 | 102 | |
| AK099337.1 | - | 1 - 2 | 191 | 102 | ||
More Information
Set Membership: |
|

LOC4325074
Genomic Map