Assay Details
| Gene Symbol: LOC4325006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765434.2 | XP_015620920.1 | 2 - 3 | 597 | 72 | |
| GenBank mRNA | AK068392.1 | - | 2 - 3 | 547 | 72 | |
| EU846993.1 | - | 2 - 3 | 506 | 72 | ||
More Information
Set Membership: |
|

LOC4325006
Genomic Map