Assay Details
Gene Symbol: LOC4324888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765696.2 | XP_015621182.1 | 2 - 3 | 378 | 71 | |
GenBank mRNA | AK070958.1 | - | 2 - 3 | 379 | 71 | |
AY551913.1 | - | 1 - 2 | 269 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
