Assay Details
| Gene Symbol: LOC4324888 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765696.2 | XP_015621182.1 | 5 - 6 | 628 | 63 | |
| GenBank mRNA | AK070958.1 | - | 5 - 6 | 629 | 63 | |
| AY551913.1 | - | 4 - 5 | 519 | 63 | ||
More Information
Set Membership: |
|

LOC4324888
Genomic Map