Assay Details
Gene Symbol: LOC4324888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765696.2 | XP_015621182.1 | 7 - 8 | 708 | 84 | |
GenBank mRNA | AK070958.1 | - | 7 - 8 | 709 | 84 | |
AY551913.1 | - | 6 - 7 | 599 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
