Assay Details
Gene Symbol: LOC4324979 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766903.2 | XP_015622389.1 | 8 - 9 | 1522 | 136 | |
GenBank mRNA | AK103626.1 | - | 8 - 9 | 1487 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() |
