Assay Details
| Gene Symbol: LOC4324983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791316.2 | XP_015646802.1 | 4 - 5 | 1535 | 68 | |
| GenBank mRNA | AB244276.1 | - | 4 - 5 | 1535 | 68 | |
More Information
Set Membership: |
|

LOC4324983
Genomic Map