Assay Details
| Gene Symbol: LOC4327751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760575.2 | XP_015616061.1 | 1 - 2 | 1336 | 62 | |
| GenBank mRNA | AK105517.1 | - | 1 - 2 | 1040 | 62 | |
More Information
Set Membership: |
|

LOC4327751
Genomic Map