Assay Details
| Gene Symbol: LOC4326948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760270.2 | 2 - 3 | 246 | 64 | ||
| GenBank mRNA | AK070499.1 | - | 2 - 3 | 204 | 64 | |
| AK119221.1 | - | 2 - 3 | 205 | 64 | ||
More Information
Set Membership: |
|

LOC4326948
Genomic Map