Assay Details
| Gene Symbol: LOC4326948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760270.2 | 6 - 7 | 538 | 64 | ||
| GenBank mRNA | AK070499.1 | - | 6 - 7 | 496 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4326948| Gene Symbol: LOC4326948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760270.2 | 6 - 7 | 538 | 64 | ||
| GenBank mRNA | AK070499.1 | - | 6 - 7 | 496 | 64 | |
Set Membership: |
|