Assay Details
| Gene Symbol: LOC4326948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760270.2 | 8 - 9 | 916 | 104 | ||
| GenBank mRNA | AK070499.1 | - | 8 - 9 | 874 | 104 | |
| AK119221.1 | - | 8 - 9 | 875 | 104 | ||
More Information
Set Membership: |
|

LOC4326948
Genomic Map