Assay Details
| Gene Symbol: LOC4326815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765771.2 | XP_015621257.1 | 10 - 11 | 1430 | 63 | |
| XM_015765772.2 | XP_015621258.1 | 10 - 11 | 1339 | 63 | ||
| GenBank mRNA | AK068780.1 | - | 10 - 11 | 1343 | 63 | |
More Information
Set Membership: |
|

LOC4326815
Genomic Map