Assay Details
| Gene Symbol: LOC4326815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765771.2 | XP_015621257.1 | 11 - 12 | 1569 | 75 | |
| XM_015765772.2 | XP_015621258.1 | 11 - 12 | 1478 | 75 | ||
| GenBank mRNA | AK059213.1 | - | 11 - 12 | 1329 | 75 | |
| AK068780.1 | - | 11 - 12 | 1482 | 75 | ||
More Information
Set Membership: |
|

LOC4326815
Genomic Map