Assay Details
| Gene Symbol: LOC4326815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765771.2 | XP_015621257.1 | 2 - 3 | 587 | 117 | |
| GenBank mRNA | AK059213.1 | - | 2 - 3 | 347 | 117 | |
| AK068780.1 | - | 2 - 3 | 500 | 117 | ||
More Information
Set Membership: |
|

LOC4326815
Genomic Map