Assay Details
| Gene Symbol: LOC4326818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766289.2 | XP_015621775.1 | 2 - 3 | 562 | 106 | |
| GenBank mRNA | AK105048.1 | - | 2 - 3 | 537 | 106 | |
More Information
Set Membership: |
|

LOC4326818
Genomic Map