Assay Details
| Gene Symbol: LOC4326818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766289.2 | XP_015621775.1 | 3 - 4 | 637 | 80 | |
| XM_015766290.2 | XP_015621776.1 | 2 - 3 | 1473 | 80 | ||
| GenBank mRNA | AK064797.1 | - | 2 - 3 | 1473 | 80 | |
| AK105048.1 | - | 3 - 4 | 612 | 80 | ||
More Information
Set Membership: |
|

LOC4326818
Genomic Map