Assay Details
| Gene Symbol: LOC4326818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766289.2 | XP_015621775.1 | 5 - 6 | 985 | 133 | |
| XM_015766290.2 | XP_015621776.1 | 4 - 5 | 1821 | 133 | ||
| GenBank mRNA | AK064797.1 | - | 4 - 5 | 1821 | 133 | |
| AK105048.1 | - | 5 - 6 | 960 | 133 | ||
More Information
Set Membership: |
|

LOC4326818
Genomic Map