Assay Details
| Gene Symbol: LOC4326524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761305.2 | XP_015616791.1 | 1 - 2 | 283 | 101 | |
| GenBank mRNA | AK101890.1 | - | 1 - 2 | 275 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015761305.2 transcripts.
Set Membership: |
|

LOC4326524
Genomic Map