Assay Details
| Gene Symbol: LOC4326538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755219.2 | XP_015610705.1 | 1 - 2 | 543 | 81 | |
| GenBank mRNA | AK099501.1 | - | 1 - 2 | 423 | 81 | |
More Information
Set Membership: |
|

LOC4326538
Genomic Map