Assay Details
Gene Symbol: LOC4326538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755219.2 | XP_015610705.1 | 2 - 3 | 620 | 87 | |
GenBank mRNA | AK099501.1 | - | 2 - 3 | 500 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
