Assay Details
| Gene Symbol: LOC4326538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755219.2 | XP_015610705.1 | 6 - 7 | 1069 | 153 | |
| GenBank mRNA | AK099501.1 | - | 6 - 7 | 949 | 153 | |
More Information
Set Membership: |
|

LOC4326538
Genomic Map