Assay Details
Gene Symbol: LOC4325393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794381.2 | XP_015649867.1 | 12 - 13 | 1779 | 156 | |
XM_015794391.2 | XP_015649877.1 | 12 - 13 | 1611 | 156 | ||
GenBank mRNA | AK070111.1 | - | 12 - 13 | 1723 | 156 | |
AK103319.1 | - | 12 - 13 | 1758 | 156 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
