Assay Details
| Gene Symbol: LOC4325393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794381.2 | XP_015649867.1 | 13 - 14 | 1942 | 70 | |
| XM_015794391.2 | XP_015649877.1 | 13 - 14 | 1774 | 70 | ||
| GenBank mRNA | AK070111.1 | - | 13 - 14 | 1886 | 70 | |
| AK103319.1 | - | 13 - 14 | 1921 | 70 | ||
More Information
Set Membership: |
|

LOC4325393
Genomic Map